Learn More
Invitrogen™ PHYKPL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113852
Description
Immunogen sequence: KIKHPIVGDV RGVGLFIGVD LIKDEATRTP ATEEAAYLVS RLKENYVLLS For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 92%; Dog: 100%; Guinea Pig: 85%; Horse: 92%; Human: 100%; Mouse: 85%; Pig: 92%; Rabbit: 92%; Rat: 92%.
This is a nuclear gene encoding a mitochondrial enzyme that catalyzes the conversion of 5-phosphonooxy-L-lysine to ammonia, inorganic phosphate, and 2-aminoadipate semialdehyde. Mutations in this gene may cause phosphohydroxylysinuria. Alternative splicing results in multiple transcript variants.
Specifications
PHYKPL | |
Polyclonal | |
Unconjugated | |
PHYKPL | |
2900006B13Rik; 5-phosphohydroxy-L-lysine phospholyase; 5-phosphohydroxy-L-lysine phospho-lyase; 5-phosphonooxy-L-lysine phospho-lyase; Agxt2l2; Alanine--glyoxylate aminotransferase 2-like 2; alanine-glyoxylate aminotransferase 2-like 2; PHLU; PHYKPL; PP9286 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
85007 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8IUZ5 | |
PHYKPL | |
Synthetic peptide directed towards the C terminal region of human AGXT2L2 (aa 351-400). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.