Description
- Example of low-grade metamorphism from easily recognizable parent rock
- For use in classroom identification labs and exercises
- Correlates well to Next Generation Science Standards Evidence Statements
- Fissile property of slate is retained, but has attained glossy sheen and soapy feel more indicative of metamorphic material
Specifications
Specifications
| Product Type | Metamorphic Rock |
| For Use With (Application) | Demonstration of metamorphic properties, such as glossy sheen and dense, foliated minerals |
| Packaging | Clear labeled and barcoded bag |
| Quantity | 12/Pk. |
| Specimen Size | 2 to 3 cm |
| Specimen Type | Phyllite |