Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Phyllite Specimen
SCI_ED

Catalog No. S26681 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S26681 1 Ea. 7 to 8 cm
S24788 1 Ea. 2 to 3 cm
S26637 10/Pk. 2 to 3 cm
S24789 12/Pk. 2 to 3 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S26681 Supplier Eisco™ Supplier No. ESNG0060HAND
Add to Cart
Add to Cart

Phyllite Specimen for demonstrations of metamorphic properties, such as glossy sheen and dense, foliated minerals.

  • Example of low-grade metamorphism from easily recognizable parent rock
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Fissile property of slate is retained, but has attained glossy sheen and soapy feel more indicative of metamorphic material

Specifications

Product Type Metamorphic Rock
Quantity 1 Ea.
Specimen Size 7 to 8 cm
Weight (Metric) 0.5 kg
Specimen Type Phyllite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.