Learn More
Eisco™ Physical Properties of Minerals Kit
SCI_ED
Catalog No. S24661 Shop All Eisco Products
Change view
Click to view available options
Quantity:
12 Specimens
Catalog No. Quantity
S24661 12 Specimens
1 options
Catalog No. S24661 Supplier Eisco™ Supplier No. ESNG0002
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
For study of color, luster, hardness, crystal habit, streak, and specific density. The Physical Properties of Minerals Kit contains 12 specimens and key.
  • For classroom and personal use
  • Numbered and stored in sturdy, sectioned container for easy access and safety
  • Average specimen size: 2 to 3 cm
  • Earth science students can explore rhombic cleavage of calcite, compare hardness of quartz and graphite, study pyrite's metallic luster
Specifications
Product Type Physical Properties of Minerals Kit
For Use With (Application) Study color, luster, hardness, crystal habit, streak, and specific density of each sample
Includes Hematite, magnetite, biotite, gypsum, calcite, hornblende, augite, labradorite, microcline, muscovite, olivine, and quartz
Packaging Sectioned container
Quantity 12 Specimens
Specimen Size Approx. 1 in.