Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Physics with Vernier
SDPSCI_ED

Catalog No. s16046nd Shop All Vernier Software Products
Change view
Click to view available options
:
contains 35 experiments
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S16046ND contains 35 experiments
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16046ND Supplier Vernier Supplier No. PWV
Only null left
Add to Cart
Add to Cart

Contains experiments that aid in learning physics concepts.

  • Contains 35 experiments in mechanics, sound, light, electricity, and magnetism
  • Enhanced graphics with experiments for more Vernier sensors and accessories
  • Includes materials list, step-by-step instructions, data tables, and questions
  • Contains Teacher Information section with directions for setting up experiments, helpful hints, and sample graphs and data
  • Word-processing files of student sections on CD
  • Recommended for College and High School

Specifications

For Use With (Application) Experiments in mechanics, sound, light, electricity, and magnetism
Product Type Lab Book
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.