Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PI 103 hydrochloride
GREENER_CHOICE

Catalog No. 29301
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-301 1 mg
29-301-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 29-301 Supplier Tocris Bioscience™ Supplier No. 2930/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inhibitor of PI 3-kinase, mTOR and DNA-PK

Inhibitor of DNA-PK, PI 3-kinase (p110α) and mTOR (IC50 values are 2, 8, 20, 26, 48, 83, 88, 150, 850, 920, ∽ 1000 and 2300 nM for DNA-PK, p110α, mTORC1, PI 3-KC2β, p110δ, mTORC2, p110β, p110γ, ATR, ATM, PI 3-KC2α and hsVPS34 respectively). Inhibits growth of human tumor xenografts in mice in vivo. Induces autophagosome formation in glioma cells.

Chemical Identifiers

CAS 371935-79-4
Molecular Formula C19H17ClN4O3

Specifications

Quantity 1 mg
Formula Weight 384.82
Purity >98%
Molecular Weight (g/mol) 384.82
Chemical Name or Material PI 103 hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.