Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PI 3-Kinase C2 beta Antibody (OTI3B1), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273395R
Description
PI 3-Kinase C2 beta Monoclonal specifically detects PI 3-Kinase C2 beta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PI 3-Kinase C2 beta | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PIK3C2B | |
Full length recombinant protein of human PI 3-Kinase C2 beta (NP_002637) produced in HEK293T cells. | |
0.1 mL | |
5287.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3B1 | |
Western Blot, Immunohistochemistry | |
C2-PI3KDKFZp686G16234, EC 2.7.1, EC 2.7.1.154, phosphatidylinositol 3-kinase C2 domain-containing beta polypeptide, phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit beta, Phosphoinositide 3-kinase-C2-beta, phosphoinositide-3-kinase, class 2, beta polypeptide, PI3K-C2beta, PI3K-C2-beta, PTDINS-3-kinase C2 beta, PtdIns-3-kinase C2 subunit beta | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction