Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PI 3-Kinase p110 beta/PIK3CB Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299597APC
Description
PI 3-Kinase p110 beta/PIK3CB Polyclonal antibody specifically detects PI 3-Kinase p110 beta/PIK3CB in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| PI 3-Kinase p110 beta/PIK3CB | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| P110BETA, PI3K, PI3KBETA, PIK3C1, PIK3CB phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit beta | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PI 3-Kinase p110 beta/PIK3CB. | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Cycle and Replication, mTOR Pathway | |
| 5291 | |
| Store at 4C in the dark. | |
| IgG |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction