Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PI 3-Kinase p85 alpha Antibody (M253), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP323296
Description
PI 3-Kinase p85 alpha Monoclonal antibody specifically detects PI 3-Kinase p85 alpha in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
PI 3-Kinase p85 alpha | |
Monoclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Chicken | |
Purified |
Western Blot, Immunofluorescence | |
M253 | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
AGM7, GRB1, IMD36, p85, p85-ALPHA, Phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha, phosphatidylinositol 3-kinase regulatory subunit alpha, phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha), phosphatidylinositol 3-kinase-associated p-85 alpha, phosphoinositide-3-kinase regulatory subunit alpha, phosphoinositide-3-kinase, regulatory subunit 1 (alpha), PI3K regulatory subunit alpha, PI3-kinase regulatory subunit alpha, PI3-kinase subunit p85-alpha, PIK3R1, PtdIns-3-kinase regulatory subunit alpha, PtdIns-3-kinase regulatory subunit p85-alpha | |
Clone M253 was generated from a recombinant protein corresponding to amino acids in the C-terminal region of human PI 3-Kinase p85 alpha. This region is homologous to similar regions in rat and mouse PI 3-Kinase p85 alpha. [UniProt# P27986] | |
0.1 mL | |
Cancer, Diabetes Research, Immune Dysfunction, Lipid and Metabolism, mTOR Pathway, Protein Kinase, Signal Transduction | |
5295 | |
Store at -20°C. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction