Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PI 3-Kinase p85 beta Antibody (T15) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $560.50
Specifications
Antigen | PI 3-Kinase p85 beta |
---|---|
Clone | T15 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Immunohistochemistry (Frozen), Western Blot, Immunoprecipitation, Immunohistochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017733
![]() |
Novus Biologicals
NB100655900.025MG |
0.025 mg |
Each for $243.50
|
|
|||||
NB10065590
![]() |
Novus Biologicals
NB10065590 |
0.1 mg |
Each for $560.50
|
|
|||||
Description
PI 3-Kinase p85 beta Monoclonal specifically detects PI 3-Kinase p85 beta in Human, Mouse, Rat, Bovine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
PI 3-Kinase p85 beta | |
1.0 mg/mL | |
Immunohistochemistry (Frozen), Western Blot, Immunoprecipitation, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Autophagy, Signal Transduction | |
P23726 | |
5296 | |
Balculovirus expressed (Recombinant) Bovine P85 beta subunit of P13 kinase. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
T15 | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Bovine, Primate, Mouse, Rat, Human | |
P85B, phosphatidylinositol 3-kinase 85 kDa regulatory subunit beta, phosphatidylinositol 3-kinase regulatory subunit beta, phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 2 (p85 beta), phosphoinositide-3-kinase, regulatory subunit 2 (beta), phosphoinositide-3-kinase, regulatory subunit 2 (p85 beta), phosphoinositide-3-kinase, regulatory subunit, polypeptide 2 (p85 beta), PI3K regulatory subunit beta, PI3-kinase regulatory subunit beta, PI3-kinase subunit p85-beta, ptdIns-3-kinase regulatory subunit beta, ptdIns-3-kinase regulatory subunit p85-beta | |
PIK3R2 | |
IgG1 | |
Protein A or G purified | |
Recognizes the p85 subunit of bovine PI-3 kinase. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title