Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega PI:3PS Lipid Kinase Substrate, 0.5mg, 0.5ml

Catalog No. PRV1711 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV1711 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV1711 Supplier Promega Supplier No. V1711
Add to Cart
Add to Cart

The lipid kinase substrates PIP2:3PS and PI:3PS are standalone substrates that are also part of the PI3K-Glo kinase profiling system.

  • Phosphoinositide−3 KInase (PI3K) Substrates
  • Use in screening assays with purified PI3K Class I lipid kinases and ADP−Glo™ Kinase Assay
  • PIP2:3PS supplied at 0.25mg in 0.25ml
  • PI:3PS supplied at 0.5mg in 0.5ml

Specifications

Quantity 0.5 mL
Content And Storage <-65°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.