Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PI 4 Kinase type 2 beta Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297902JF549
Description
PI 4 Kinase type 2 beta Polyclonal specifically detects PI 4 Kinase type 2 beta in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
PI 4 Kinase type 2 beta | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
EC 2.7.1.67, FLJ11105, phosphatidylinositol 4-kinase type 2 beta, phosphatidylinositol 4-kinase type 2-beta, Phosphatidylinositol 4-kinase type II-beta, phosphatidylinositol 4-kinase type-II beta, PI4KIIB, PI4KII-BETA, PIK42B | |
Produced in rabbits immunized with E. coli-derived Human PI 4 Kinase type 2 beta fragment. | |
0.1 mL | |
Protein Kinase | |
55300 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction