Learn More
Invitrogen™ PI(5)P Lipid Kinase Substrate
The PI(5)P Lipid Kinase Substrate has been specifically optimized for use with Type II PIP kinases (e.g., PIP4K2A)
$1198.00 - $8930.00
Specifications
Content And Storage | Phosphatidylinositol 5-phosphate (PI(5)P) in 30 mM HEPES pH 7.5 buffer. |
---|---|
Form | Powder |
Product Line | Invitrogen™ |
Substrate | Phosphatidylinositol 5-phosphate (PI(5)P) |
Substrate Type | Kinase Substrate |
Description
- The PI(5)P Lipid Kinase Substrate has been specifically optimized for use with Type II PIP kinases (e.g., PIP4K2A)
- The substrate consists of solubilized phosphatidylinositol 5-phosphate (PI(5)P) in an aqueous buffer, and is used in combination with our Adapta™ Universal Kinase Assay to help identify unique kinase inhibitors.
- Validated for use with Thermo Fisher Scientific lipid kinases • Available in multiple pack sizes for convenient use
- Also available in our SelectScreen™ Kinase Profiling Service
Specifications
Phosphatidylinositol 5-phosphate (PI(5)P) in 30 mM HEPES pH 7.5 buffer. | |
Invitrogen™ | |
Kinase Substrate |
Powder | |
Phosphatidylinositol 5-phosphate (PI(5)P) | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.