Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ PI 828
GREENER_CHOICESDP

Catalog No. 28141 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-141 1 mg
28-145-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 28-141 Supplier R&D Systems™ Supplier No. 2814/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. PI 3-kinase inhibitor, more potent than LY 294002 (Cat. No. 1130)

PI 3-Kinase inhibitor (IC50 values are 0.098, 0.183, 0.227 and 1.967 μM for p110β, p110α, p110δ and p110γ respectively) that displays higher potency than LY 294002 (Cat No. 1130). Can be immobilized onto a solid phase.

Chemical Identifiers

CAS 942289-87-4
Molecular Formula C19H18N2O3

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 322.36
Purity >99%
Molecular Weight (g/mol) 322.364
Chemical Name or Material PI 828
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.