Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva pI Markers

Catalog No. 45000083 Shop All Cytiva Products
Change view
Click to view available options
pH Range:
3 to 10
5 to 10.5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
45-000-083 5 to 10.5
45-000-081 3 to 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-000-083 Supplier Cytiva Supplier No. 17047301
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Run these markers in parallel with the experimental samples on isoelectric focusing gels. Cytiva Life Sciences™ pI Markers are lyophilized mixtures of stable, salt-free, highly purified proteins.

The markers may be run in parallel with the experimental samples on isoelectric focusing gels and are not prestained.

  • The precisely defined pI of the markers allows a permanent and accurate calibration of sample components
  • pH gradients and pI can be measured to within 0.01 to 0.05 pH unit
  • The progress of the focusing experiment can be monitored visually by the focusing of methyl red or Cytochrome c

Not for use in denaturing (urea) systems.

TRUSTED_SUSTAINABILITY

Specifications

Color White
pH Range 5 to 10.5
Form Lyophilized
For Use With (Application) To be used for isoelectric focusing (IEF)
Kit Contents b-Lactoglobulin A - 5.20 pI (Native), Carbonic Anhydrase B (Bovine) - 5.85 pI (Native), Carbonic Anhydrase B (human) - 6.55 pI (Native), Myoglobin, Acidic - 6.85 pI (Native), Myoglobin, basic - 7.35 pI (Native), Lentil Lectin, Acidic - 8.15 pI (Native)
Quantity 1 Kit: 10 Vials
Range 5.2 to 10.25 pl at 24°C
Storage Requirements 4°C
Product Type pl Marker Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.