Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ PI/RNAse
SDP

Catalog No. 48176004 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-176-004 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 48-176-004 Supplier R&D Systems™ Supplier No. 48176004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

R&D Systems™ PI/RNAse can be added directly to the sample just prior to flow cytometry.

  • Propidium iodide/RNAse solution
  • Typically 10μL per 500μL sample is sufficient
  • Incubate at room temperature for 5 minutes then analyze
  • If propidium iodide is used perform each of the above controls in duplicate so that the samples can be assayed with and without propidium iodide
  • 1mg/mL propidium iodide; 50mg/mL RNase A

Specifications

Content And Storage Store the unopened product at 2 to 8°C
Quantity 1 mL
Product Line R&D Systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.