Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PI16 Monoclonal Antibody (5D4C7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548813
Description
PI16 Monoclonal Antibody for Western Blot, Flow, ELISA
The PI16 gene encodes the protein peptidase inhibitor 16, which is a member of the protease inhibitor family and plays roles in modulating protease activity within tissues. PI16 is primarily expressed in the prostate but is also found in other tissues, where it contributes to extracellular matrix organization and integrity by inhibiting serine proteases. By regulating protease activity, PI16 helps maintain tissue homeostasis and influences processes such as cell proliferation, differentiation, and migration. This protein's function is significant in preventing excessive proteolytic activity that could lead to tissue damage or contribute to pathological conditions like inflammation and cancer. Specifically, PI16's role in the prostate gland has been associated with modulating growth and repair mechanisms, providing potential implications in prostate cancer progression where protease activity is often dysregulated. Current research focuses on understanding PI16's interactions and regulatory mechanisms to explore its therapeutic potential in diseases characterized by abnormal protease activities.
Specifications
PI16 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q6UXB8 | |
PI16 | |
Purified recombinant fragment of human CD364 (AA: 79-210) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
5D4C7 | |
Unconjugated | |
PI16 | |
1200009H11Rik; CD364; Cripi; CRISP9; CRISP-9; Cysteine-rich protease inhibitor; cysteine-rich secretory protein 9; microseminoprotein, beta-binding protein; MSMBBP; peptidase inhibitor 16; Pi16; PI-16; protease inhibitor 16; PSEC0164; PSP94-binding protein; PSPBP; UNQ289/PRO328 | |
Mouse | |
Protein G | |
RUO | |
221476 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction