Learn More
Description
Specifications
Specifications
| Antigen | PI3 Kinase |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | Please refer to lot specific datasheet. |
| Conjugate | Unconjugated |
| Gene Symbols | Pik3r1 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated linear peptide corresponding to 18 amino acids from the C-terminal region of p85 regulatory subunit of PI-3 kinase alpha. |
| Purification Method | Affinity Purified |
| Quantity | 200 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.