Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI3 Kinase Rabbit anti-Rat, Human, Agarose, Polyclonal, MilliporeSigma™

Catalog No. 16107 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16107 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16107 Supplier MilliporeSigma Supplier No. 16107
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-PI3 Kinase Antibody, p85, agarose conjugate detects level of PI3 Kinase & has been published & validated for use in IP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI3 Kinase
Applications Immunoprecipitation
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Agarose
Formulation PBS, pH 7.4, containing 0.05% sodium azide
Gene Symbols PIK3R1;GRB1;PI3K;p85-ALPHA
Host Species Rabbit
Quantity 200 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NM_181504.2;NM_181523.1;NM_181524.1
Target Species Rat, Human
Content And Storage 1 year at 4°C
Product Type Antibodies
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.