Learn More
Promega PI3K-Glo Class I Profiling Kit, 1 each
Catalog No. PRV1690 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
PRV1690 1 Ea.
1 options
Catalog No. PRV1690 Supplier Promega Supplier No. V1690
Add to Cart
Add to Cart
Phosphatidylinositol (PI) and its phosphorylated derivates, collectively called phosphoinositides, are important second messengers that are critical as signaling molecules and for cellular membrane remodeling.
  • Simple, Add−and−Read Assay to Detect Class I PI3K Activity
  • Includes purified recombinant proteins of class I PI3Ks, optimized reaction buffer, ready−to−use lipid kinase substrates and ADP−Glo™ Kinase Assay
  • High signal−to−background ratios even at low percent substrate conversion
  • Easily scalable to 1,536−well format
Specifications
Quantity 1 Ea.
Content And Storage <-65°C