Learn More
Promega PI3K (p110alpha[H1047R]/p85alpha), 20ug, 200microl
Catalog No. PRV1741 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
200 μL
Catalog No. Quantity
PRV1741 200 μL
1 options
Catalog No. PRV1741 Supplier Promega Supplier No. V1741
The PI3K lipid kinase enzymes are used with ADP-Glo KInase Assay System and Lipid Kinase Substrates in a homogeneous, add-mix-read lipid kinase assay.
  • Purified PI3K Class I Lipid Kinases
  • Use with ADP−Glo™ Assay and PI3K Substrates in lipid kinase activity assays
Specifications
Quantity 200 μL
Content And Storage <-65°C