Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PI3K p55 gamma Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110171
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110171 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110171 Supplier Invitrogen™ Supplier No. PA5110171
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PDLIQLRKIR DQHLVWLNHK GVRQKRLNVW LGIKNEDADE NYFINEEDEN LPHYDEKTWF VEDINRVQAE DLLYGKPDGA FLIRESSKK.

PIK3R3 binds to activated (phosphorylated) protein-tyrosine kinases through its SH2 domain and regulates their kinase activity. During insulin stimulation, it also binds to IRS-1. It is a component of a heterodimer of p110 (catalytic) and p55 (regulatory) subunits. The protein is expressed at highest levels in brain and testis. Lower levels are detected in adipose tissue, kidney, heart, lung and skeletal muscle. The protein contains 2 SH2 domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI3K p55 gamma
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.278 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Pik3r3
Gene Accession No. Q63789, Q64143, Q92569
Gene Alias AA414954; p55; p55-GAMMA; p55pik; phosphatidylinositol 3 kinase, regulatory subunit, polypeptide 3 (p55); phosphatidylinositol 3-kinase 55 kDa regulatory subunit gamma; phosphatidylinositol 3-kinase p55 subunit; phosphatidylinositol 3-kinase regulatory subunit gamma; phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 3 (p55, gamma); phosphoinositide-3-kinase regulatory subunit 3; phosphoinositide-3-kinase, regulatory subunit 3 (gamma); PI3K regulatory subunit gamma; PI3-kinase p85 subunit gamma; PI3-kinase regulatory subunit gamma; PI3-kinase subunit p55-gamma; Pik3r3; ptdIns-3-kinase p85-gamma; ptdIns-3-kinase regulatory subunit gamma; ptdIns-3-kinase regulatory subunit p55-gamma; RP4-533D7.2
Gene Symbols Pik3r3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 300-388 of human PIK3R3/p55PIK (NP_0036203).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18710, 60664, 8503
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.