Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PI3K p85 alpha Monoclonal Antibody (A3-D0)
GREENER_CHOICE

Catalog No. PIMA532917
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532917 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532917 Supplier Invitrogen™ Supplier No. MA532917
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PI3K p85 alpha Monoclonal Antibody for ICC/IF, Western Blot

Phosphatidylinositol 3-kinase (PIK3) is a signaling protein that phosphorylates the inositol ring of phosphatidylinositol at the 3-prime position. Signaling is initiated when upstream tyrosine kinase receptors or G protein coupled receptors are bound by their ligands. It binds to activated (phosphorylated) protein-Tyr kinases, through its SH2 domain, and acts as an adapter, mediating the association of the p110 catalytic unit to the plasma membrane. Phosphatidylinositol 3-kinase plays an important role in the metabolic actions of insulin, and a mutation in this gene has been associated with insulin resistance. During insulin stimulation, it also binds to IRS-1. Mutations in the gene can result in Agammaglobulinemia 7, SHORT syndrome, or Immunodeficiency 36.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI3K p85 alpha
Applications Western Blot, Immunocytochemistry, Immunohistochemistry, Western Blot, Western Blot
Classification Monoclonal
Clone A3-D0
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene PIK3R1
Gene Accession No. P27986
Gene Alias AGM7; GRB1; IMD36; p50alpha; p55alpha; p55-GAMMA; p55PIK; p85; p85alpha; p85-ALPHA; phosphatidylinositol 3-kinase; phosphatidylinositol 3-kinase 55 kDa regulatory subunit gamma; phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha; Phosphatidylinositol 3-kinase regulatory subunit alpha; phosphatidylinositol 3-kinase regulatory subunit gamma; phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha); phosphatidylinositol 3-kinase-associated p-85 alpha; Phosphoinositide 3-kinase p85 (other splicing variants: p55 and p50); Phosphoinositide 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha); phosphoinositide-3-kinase regulatory subunit; phosphoinositide-3-kinase regulatory subunit 1; phosphoinositide-3-kinase regulatory subunit alpha; phosphoinositide-3-kinase, regulatory subunit 1 (alpha); phosphoinositide-3-kinase, regulatory subunit 1 (p85 alpha); phosphoinositide-3-kinase, regulatory subunit, polypeptide 1 (p85 alpha); PI3K; PI3K regulatory subunit alpha; PI3K regulatory subunit gamma; PI3KA; PI3-kinase p85 subunit alpha; PI3-kinase p85-subunit alpha; PI3-kinase regulatory subunit alpha; PI3-kinase regulatory subunit gamma; PI3-kinase subunit p55-gamma; PI3-kinase subunit p85-alpha; Pik3r1; polypeptide 1 p85 alpha; ptdIns-3-kinase p85-alpha; ptdIns-3-kinase regulatory subunit alpha; PtdIns-3-kinase regulatory subunit p85-al; ptdIns-3-kinase regulatory subunit p85-alpha; RP4-533D7.2
Gene Symbols PIK3R1
Host Species Mouse
Immunogen Recombinant protein within Human PI3-kinase p85 subunit alpha aa 19-219.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5295
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.