Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI3KCA Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984485
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4485 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4485 Supplier Bioss Supplier No. BS2067R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phosphatidylinositol 3-kinase is composed of an 85 kDa regulatory subunit and a 110 kDa catalytic subunit. The protein encoded by this gene represents the catalytic subunit, which uses ATP to phosphorylate PtdIns, PtdIns4P and PtdIns(4,5)P2. This gene has been found to be oncogenic and has been implicated in cervical cancers.

Specifications

Antigen PI3KCA
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PIK3CA
Gene Accession No. P42336
Gene Alias 2.7.1.153; 6330412C24Rik; BOS_1038; caPI3K; CLOVE; CWS5; EC:2.7.11.1; GRB1; MCAP; MCM; MCMTC; MGC142161; MGC142163; p110; p110alpha; p110-alpha; phosphatidylinositol 3-kinase, catalytic, 110-KD, alpha; phosphatidylinositol 3-kinase, catalytic, alpha polypeptide; Phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit, alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit alpha; Phosphoinositide-3-kinase catalytic alpha polypeptide; phosphoinositide-3-kinase, catalytic, alpha polypeptide; PI3K; PI3K alpha; PI3Kalpha; PI3K-alpha; PI3-kinase p110 subunit alpha; PI3-kinase subunit alpha; Pik3ca; PIK3R1; ptdIns-3-kinase p110; ptdIns-3-kinase subunit alpha; PtdIns-3-kinase subunit p110-alpha; Serine/threonine protein kinase PIK3CA
Gene Symbols PIK3CA
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PI3KCA .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5290
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.