Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI4K alpha, type IRabbit anti-Human, Polyclonal, Rockland™
SDP

Catalog No. 501992275
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2275 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-2275 Supplier Rockland Immunochemicals Supplier No. 600401J91
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. A BLAST analysis was used to suggest cross-reactivity with mouse and human based on 100% sequence homology. Cross-reactivity with PI4K, type I alpha from other sources has not been determined.

Overexpression of phosphatidylinositol phosphate 5-kinase alpha (PIP5KIalpha), which synthesizes PIP2, suppresses apoptosis, whereas a kinase-deficient mutant does not. Protection by the wild-type PIP5KIalpha isaccompanied by decreases in the generation of activated caspases and of caspase 3-cleaved PARP. Protection is not mediated through PIP3 or Akt activation. An anti-apoptotic role for PIP(2) is substantiated by the finding that PIP5KIalpha is cleaved by caspase 3 during apoptosis, and cleavage inactivates PIP5KIalpha in vitro. Mutation of the P(4) position (D279A) of the PIP5KIalpha caspase 3 cleavage consensus prevents cleavage in vitro, and during apoptosis in vivo. Significantly, the caspase 3-resistant PIP5KIalpha mutant is more effective in suppressing apoptosis than the wild-type kinase. PIP2 is a direct regulator of apical and effector caspases in the death receptor and mitochondrial pathways, and PIP5KIalpha inactivation contributes to the progression of apoptosis.

Specifications

Antigen PI4K alpha, type I
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl, 30% glycerol and no preservative; pH 7.2
Gene Pip5k1a
Gene Accession No. Q99755
Gene Alias 68 kDa type I phosphatidylinositol 4-phosphate 5-kinase; 68 kDa type I phosphatidylinositol 4-phosphate 5-kinase alpha; 68 kDa type I phosphatidylinositol-4-phosphate 5-kinase; 68 kDa type I phosphatidylinositol-4-phosphate 5-kinase alpha; BOS_2900; Phosphatidylinositol 4-phosphate 5-kinase type I alpha; Phosphatidylinositol 4-phosphate 5-kinase type I beta; Phosphatidylinositol 4-phosphate 5-kinase type-1 alpha; phosphatidylinositol-4-phosphate 5-kinase type 1 alpha; phosphatidylinositol-4-phosphate 5-kinase type I alpha; phosphatidylinositol-4-phosphate 5-kinase type-1 alpha; phosphatidylinositol-4-phosphate 5-kinase, type 1 alpha; phosphatidylinositol-4-phosphate 5-kinase, type 1 beta; phosphatidylinositol-4-phosphate 5-kinase, type 1, alpha; phosphatidylinositol-4-phosphate 5-kinase, type I, alpha; PI4P5K-I[a]; PI4P5KIbeta; PI51A; Pip5k1a; PIP5K1-alpha; Pip5k1b; PIP5KIalpha; Pipk5a; ptdIns(4)P-5-kinase 1 alpha
Gene Symbols Pip5k1a
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8394
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.