Learn More
Invitrogen™ PI4K2A Monoclonal Antibody (3E1)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA522144
Catalog No. PIMA522144
Only null left
Add to Cart
Description
Description
Peptide Sequence: LILPKISDPN FVKDLEEDLY ELFKKDPGFD RGQFHKQIAV MRGQILNLTQ ALKDNKSPLH LVQMPPVIVE TARSHQRSSS ESYTQSFQSR KPFFS.
PI4K2A (PI4K2 alpha) lipid kinase or phosphatidylinositol 4-kinase type 2 alpha is involved in the formation of phosphatidylinositolpolyphosphates (PtdInsPs) which are centrally involved in many biologic processes ranging from cell growth and organization of the actin cytoskeleton to endo- and exocytosis. PI4K2A phosphorylates PtdIns at the D-4 position which is an essential step in the biosynthesis of PtdInsPs. PI4K2A is a novel regulator of tumor growth by its action on angiogenesis and HIF-1alpha regulation. PI4K2A generates PtdIns4P-rich domains within the Golgi which regulate targeting of clathrin adaptor AP-1 complexes. PI4K2A is also required for Wnt to regulate LRP6 phosphorylation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PI4K2A
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.4
Q9BTU6
Pi4k2a
PI4K2A (NP_060895.1, 383 a.a. ~ 477 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2b κ
ELISA
3E1
Unconjugated
Pi4k2a
55 kDa type II phosphatidylinositol 4-kinase; null; phosphatidylinositol 4-kinase; phosphatidylinositol 4-kinase type 2 alpha; phosphatidylinositol 4-kinase type 2-alpha; phosphatidylinositol 4-kinase type II; phosphatidylinositol 4-kinase type II (PI4KII); phosphatidylinositol 4-kinase type II-alpha; Pi4k2; PI4K2A; Pi4kii; PIK42A
Mouse
Affinity Chromatography
RUO
55361
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.