Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PI4K2A (PI4K2 alpha), GST Tag Recombinant Protein

Catalog No. A32905
Encompass_Preferred
Click to view available options
Quantity:
1 mg
100 μg
20 μg

Recombinant Protein

PI4K2A (PI4K2 alpha) lipid kinase or phosphatidylinositol 4-kinase type 2 alpha is involved in the formation of phosphatidylinositolpolyphosphates (PtdInsPs) which are centrally involved in many biologic processes ranging from cell growth and organization of the actin cytoskeleton to endo- and exocytosis. PI4K2A phosphorylates PtdIns at the D-4 position which is an essential step in the biosynthesis of PtdInsPs. PI4K2A is a novel regulator of tumor growth by its action on angiogenesis and HIF-1alpha regulation. PI4K2A generates PtdIns4P-rich domains within the Golgi which regulate targeting of clathrin adaptor AP-1 complexes. PI4K2A is also required for Wnt to regulate LRP6 phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BTU6
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 55361
Molecular Weight (g/mol) 80.3 kDa
Name Human PI4K2A (PI4K2 alpha), GST Tag
Quantity 100 μg
Gene Alias 55 kDa type II phosphatidylinositol 4-kinase; null; phosphatidylinositol 4-kinase; phosphatidylinositol 4-kinase type 2 alpha; phosphatidylinositol 4-kinase type 2-alpha; phosphatidylinositol 4-kinase type II; phosphatidylinositol 4-kinase type II (PI4KII); phosphatidylinositol 4-kinase type II-alpha; Pi4k2; PI4K2A; Pi4kii; PIK42A
Common Name PI4K2A
KinaseFamily PI3 Kinase
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Full Length, Recombinant, Active
Form Liquid
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.