Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PI4K2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA515275
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA515275 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA515275 Supplier Invitrogen™ Supplier No. PA515275
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PI4K2A Polyclonal Antibody for ICC/IF, IHC (P), Western Blot

PI4K2A (PI4K2 alpha) lipid kinase or phosphatidylinositol 4-kinase type 2 alpha is involved in the formation of phosphatidylinositolpolyphosphates (PtdInsPs) which are centrally involved in many biologic processes ranging from cell growth and organization of the actin cytoskeleton to endo- and exocytosis. PI4K2A phosphorylates PtdIns at the D-4 position which is an essential step in the biosynthesis of PtdInsPs. PI4K2A is a novel regulator of tumor growth by its action on angiogenesis and HIF-1alpha regulation. PI4K2A generates PtdIns4P-rich domains within the Golgi which regulate targeting of clathrin adaptor AP-1 complexes. PI4K2A is also required for Wnt to regulate LRP6 phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI4K2A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Pi4k2a
Gene Accession No. Q2TBE6, Q9BTU6
Gene Alias 55 kDa type II phosphatidylinositol 4-kinase; null; phosphatidylinositol 4-kinase; phosphatidylinositol 4-kinase type 2 alpha; phosphatidylinositol 4-kinase type 2-alpha; phosphatidylinositol 4-kinase type II; phosphatidylinositol 4-kinase type II (PI4KII); phosphatidylinositol 4-kinase type II-alpha; Pi4k2; PI4K2A; Pi4kii; PIK42A
Gene Symbols Pi4k2a
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human PI4K2A (PI4K II).
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55361, 84095
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.