Learn More
Invitrogen™ PI4K2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595793
Description
Immunogen sequence: YQGSSGSYFV KDPQGRIIAV FKPKNEEPYG HLNPKWTKWL QKLCCPCCFG RDCLVLNQGY LSEAGASLVD QKLELNIVPR TKVVYLASET FNYSAIDRVK SRGKRLALEK VPKVGQRFNR IGLPPKVGSF QLFVEGYKDA DYWLRRFEAE PLPENTNRQL LLQFERLVVL DYIIRNTDRG NDNWLIKYDC PMDSSSSRDT DWVVVKEPVI KVAAIDNGLA FPLKHPDSWR AYPFYWAWLP QAKVPFSQEI KDLILPKISD PNFVKDLEED LYELFKKDPG FDRGQFHKQI AVMRGQILNL TQALKDNKSP LHLVQMPPVI VETARSHQRS SSESYTQSFQ SRKPFFSWW; Positive Samples: HeLa; Cellular Location: Cell junction, Cell membrane, Cell projection, Cytoplasmic vesicle, Endosome, Golgi apparatus, Lipid-anchor, Membrane, Membrane raft, Mitochondrion, dendrite, presynaptic cell membrane, synapse, synaptosome, trans-Golgi network membrane.
PI4K2A (PI4K2 alpha) lipid kinase or phosphatidylinositol 4-kinase type 2 alpha is involved in the formation of phosphatidylinositolpolyphosphates (PtdInsPs) which are centrally involved in many biologic processes ranging from cell growth and organization of the actin cytoskeleton to endo- and exocytosis. PI4K2A phosphorylates PtdIns at the D-4 position which is an essential step in the biosynthesis of PtdInsPs. PI4K2A is a novel regulator of tumor growth by its action on angiogenesis and HIF-1alpha regulation. PI4K2A generates PtdIns4P-rich domains within the Golgi which regulate targeting of clathrin adaptor AP-1 complexes. PI4K2A is also required for Wnt to regulate LRP6 phosphorylation.
Specifications
| PI4K2A | |
| Polyclonal | |
| Unconjugated | |
| Pi4k2a | |
| 55 kDa type II phosphatidylinositol 4-kinase; null; phosphatidylinositol 4-kinase; phosphatidylinositol 4-kinase type 2 alpha; phosphatidylinositol 4-kinase type 2-alpha; phosphatidylinositol 4-kinase type II; phosphatidylinositol 4-kinase type II (PI4KII); phosphatidylinositol 4-kinase type II-alpha; Pi4k2; PI4K2A; Pi4kii; PIK42A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55361 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.66 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9BTU6 | |
| Pi4k2a | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 131-479 of human PI4K2A (NP_0608951). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.