Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PI4K2B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA130306
Description
Recommended positive controls: Mouse brain and Saos 2 cell lysate.
PI4K2B lipid kinase is a member of the phosphatidylinositol 4-kinase family (PI4K) that phosphorylate phosphatidylinositol to generate phosphatidylinositol 4-phosphate which are immediate precursor of several important signaling and scaffolding molecules. PI4K2B is primarily a cytosolic PI4K that is recruited to membranes where it stimulates phosphatidylinositol 4,5-bisphosphate syntheses. PI4K2B uses phosphatidylinositol as its primary substrate and has no activity on phosphatidylinositol monophosphates. PI4K2B is a restricted minor histocompatibility antigen in patients that had been successfully treated with donor lymphocyte infusions for relapsed chronic myeloid leukemia after allogeneic stem cell transplantation. Phosphatidylinositol 4-kinases (PI4Ks) phosphorylate phosphatidylinositol to generate phosphatidylinositol 4-phosphate (PIP), an immediate precursor of several important signaling and scaffolding molecules. PIP itself may also have direct functional and structural roles. PI4K2B is a primarily cytosolic PI4K that is recruited to membranes, where it stimulates phosphatidylinositol 4, 5-bisphosphate synthesis (Wei et al., 2002 ).
Specifications
PI4K2B | |
Polyclonal | |
Unconjugated | |
PI4K2B | |
2610042N09Rik; 4933409G22Rik; hypothetical protein; phosphatidylinositol 4-kinase; phosphatidylinositol 4-kinase type 2 beta; Phosphatidylinositol 4-kinase type 2 beta; Phosphatidylinositol 4-kinase type 2-beta; phosphatidylinositol 4-kinase type 2-beta; Phosphatidylinositol 4-kinase type II-beta; phosphatidylinositol 4-kinase type-II beta; PI4K2B; PI4K2B protein; PI4KIIB; PI4KII-BETA; PIK42B; RCJMB04_25j21 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
55300, 67073 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 0.09% sodium azide | |
Q8CBQ5, Q8TCG2 | |
PI4K2B | |
KLH conjugated synthetic peptide between 305-336 amino acids from the C-terminal region of human PI4K2B. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction