Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PI4KA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572329
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572329 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572329 Supplier Invitrogen™ Supplier No. PA572329
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PI4KA Polyclonal Antibody for Western Blot

PI4KB and PI4KA are the two human type III PI4 kinases, which share sequence similarly with PI3 kinases and phosphorylate the 4 position of phosphotidyl inositol. The mammalian PI 4-kinases have been classified into two types, II and III, based on their molecular mass, and modulation by detergent and adenosine. The protein encoded by PI4KA is a type III enzyme that is not inhibited by adenosine. Two transcript variants encoding different isoforms have been described for PI4KA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI4KA
Applications Western Blot
Classification Polyclonal
Concentration 0.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PI4KA
Gene Accession No. E9Q3L2
Gene Alias leuserpin 2; phosphatidylinositol 4-kinase; phosphatidylinositol 4-kinase 230; phosphatidylinositol 4-kinase a; phosphatidylinositol 4-kinase alpha; phosphatidylinositol 4-kinase, catalytic, alpha; phosphatidylinositol 4-kinase, catalytic, alpha polypeptide; phosphatidylinositol 4-kinase, type III, alpha; pi4K230; Pi4ka; PI4K-ALPHA; PI4-kinase alpha; Pik4; PIK4CA; PMGYCHA; ptdIns-4-kinase alpha; testicular secretory protein Li 35
Gene Symbols PI4KA
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1460-1490 amino acids of mouse PI4KA.
Purification Method Protein A, Antigen affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224020
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.