Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI4KA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501727964 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7964 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7964 Supplier Proteintech Group Inc Supplier No. 124111AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a phosphatidylinositol (PI) 4-kinase which catalyzes the first committed step in the biosynthesis of phosphatidylinositol 4,5-bisphosphate. The mammalian PI 4-kinases have been classified into two types, II and III, based on their molecular mass, and modulation by detergent and adenosine. The protein encoded by this gene is a type III enzyme that is not inhibited by adenosine. Two transcript variants encoding different isoforms have been described for this gene.

Specifications

Antigen PI4KA
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PI4KA
Gene Accession No. E9Q3L2, O08662, P42356
Gene Alias PI4 kinase alpha, PI4K ALPHA, pi4K230, PI4KA, PIK4, PIK4CA, PtdIns 4 kinase alpha
Gene Symbols PI4KA
Host Species Rabbit
Immunogen PI4KA Fusion Protein Ag3062
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224020, 5297, 64161
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.