Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PI4KAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113130
Description
PI4KAP1 Polyclonal Antibody for Western Blot, ELISA
PI4KAP1 (Putative inactive phosphatidylinositol 4-kinase alpha-like protein P1) could be the product of a pseudogene. Much shorter than other members of the family and lacks the PI3K/PI4K catalytic domain.
Specifications
| PI4KAP1 | |
| Polyclonal | |
| Unconjugated | |
| PI4KAP1 | |
| phosphatidylinositol 4-kinase alpha pseudogene 1; PI4KAP1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 728233 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8N8J0 | |
| PI4KAP1 | |
| Recombinant Human Putative inactive phosphatidylinositol 4-kinase alpha-like protein P1 protein (1-262AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction