Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PI4KB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552271
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552271 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552271 Supplier Invitrogen™ Supplier No. PA552271
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TPTAFKRDPE DPSAVALKEP WQEKVRRIRE GSPYGHLPNW RLLSVIVKCG DDLRQELLAF QVLKQLQSIW EQERVPLWIK PYKILVISAD SGMIEPVVNA VSIHQVKKQS QLSLLDYFLQ EHGSYTTEAF LSAQRNFVQS CAGY Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

PI4KB and PI4KA are the two human type III PI4 kinases, which share sequence similarly with PI3 kinases and phosphorylate the 4 position of phosphotidyl inositol. Transporter for alanine, serine, cysteine, and threonine. Exhibits sodium dependence. Target protein phosphorylates phosphatidylinositol (PtdIns) at the D4 position of the inositol ring; may play a role in neuronal differentiation and maturation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI4KB
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pi4kb
Gene Accession No. Q9UBF8
Gene Alias catalytic phosphatidylinositol 4-kinase beta; ESTM41; lipid kinase; DrPI4K III beta; NPIK; phosphatidylinositol 4-kinase; phosphatidylinositol 4-kinase beta; phosphatidylinositol 4-kinase III beta; phosphatidylinositol 4-kinase, catalytic, beta; phosphatidylinositol 4-kinase, catalytic, beta polypeptide; phosphatidylinositol 4-kinase, wortmannin-sensitive; PI4K92; PI4KB; PI4Kbeta; PI4K-beta; PI4KIII; PI4KIIIBETA; PI4KIII-beta; Pik4cb; PtdIns 4-kinase beta; type III phosphatidylinositol 4-kinase beta; zgc:152715
Gene Symbols Pi4kb
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PI4KB. Recombinant protein control fragment (Product #RP-89168).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5298
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.