Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PI4KB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110166
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110166 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110166 Supplier Invitrogen™ Supplier No. PA5110166
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AFQVLKQLQS IWEQERVPLW IKPYKILVIS ADSGMIEPVV NAVSIHQVKK QSQLSLLDYF LQEHGSYTTE AFLSAQRNFV QSCAGYCLVC YLLQVKDRHN GNILLDAEGH IIHIDFGFIL SSSPRNLGFE TSAFKLTTEF VDVMGGLDGD MFNYYKMLML QGLIAARKHM DKVVQIVEIM QQGSQLPCFH GSSTIRNLKE RFHMSMTEEQ LQLLVEQMVD GSMRSITTKL YDGFQYLTNG IM.

PI4KB and PI4KA are the two human type III PI4 kinases, which share sequence similarly with PI3 kinases and phosphorylate the 4 position of phosphotidyl inositol. Transporter for alanine, serine, cysteine, and threonine. Exhibits sodium dependence. Target protein phosphorylates phosphatidylinositol (PtdIns) at the D4 position of the inositol ring; may play a role in neuronal differentiation and maturation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI4KB
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Pi4kb
Gene Accession No. Q8BKC8
Gene Alias catalytic phosphatidylinositol 4-kinase beta; ESTM41; lipid kinase; DrPI4K III beta; NPIK; phosphatidylinositol 4-kinase; phosphatidylinositol 4-kinase beta; phosphatidylinositol 4-kinase III beta; phosphatidylinositol 4-kinase, catalytic, beta; phosphatidylinositol 4-kinase, catalytic, beta polypeptide; phosphatidylinositol 4-kinase, wortmannin-sensitive; PI4K92; PI4KB; PI4Kbeta; PI4K-beta; PI4KIII; PI4KIIIBETA; PI4KIII-beta; Pik4cb; PtdIns 4-kinase beta; type III phosphatidylinositol 4-kinase beta; zgc:152715
Gene Symbols Pi4kb
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 560-801 of human PI4KB (NP_001185702).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107650
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.