Learn More
PI4KB Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303734A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Zebrafish, X. tropicalis and Bovine
Phosphorylates phosphatidylinositol (PI) in the first committed step in the production of the second messenger inositol-1, 4, 5, -trisphosphate (PIP). PI4KB may regulate Golgi disintegration/reorganization during mitosis, possibly via its phosphorylation.Specifications
PI4KB | |
Polyclonal | |
Unconjugated | |
PI4KB | |
Phosphatidylinositol 4-kinase, catalytic beta, PI4K-BETA, PI4KIIIbeta, PI4Kbeta, PIK4CB, pi4K92, NPIK, PI4K92, PI4KBETA, PI4KIIIBETA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5298 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9UBF8 | |
PI4KB | |
Between 500 and 550 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.