Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI4KB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

$518.60

Specifications

Antigen PI4KB
Concentration 0.13 mg/mL
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-172-8601
SDP
Encompass_Preferred
View Documents
Proteintech Group Inc
132471AP150UL
150 μL
Each of 1 for $518.60
Only null left
Add to Cart
 
Description

Description

Transporter for alanine, serine, cysteine, and threonine. Exhibits sodium dependence.
Specifications

Specifications

PI4KB
Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
O08561, Q8BKC8, Q9UBF8
107650, 5298, 81747
PI4KB Fusion Protein Ag3989
Primary
-20°C
PI4KB
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
PI4 kinase beta, PI4K BETA, pi4K92, PI4KB, PI4Kbeta, PI4KIIIbeta, PIK4CB, PtdIns 4 kinase beta
PI4KB
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.