Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIAS1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177158MFV610
Description
PIAS1 Polyclonal antibody specifically detects PIAS1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
PIAS1 | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
AR interacting protein, DDXBP1MGC141878, DEAD/H (Asp-Glu-Ala-Asp/His) box binding protein 1, DEAD/H box-binding protein 1, E3 SUMO-protein ligase PIAS1, GBPRNA helicase II-binding protein, GU/RH-II, Gu-binding protein, MGC141879, Protein inhibitor of activated STAT protein 1, protein inhibitor of activated STAT, 1, protein inhibitor of activated STAT-1, zinc finger, MIZ-type containing 3, ZMIZ3 | |
Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human PIAS1. The immunogen is located within the last 50 amino acids of PIAS1. Amino Acid Squence: SNSLRESHSHTVTNR | |
0.1 mL | |
Signal Transduction | |
8554 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction