Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIAS1 Recombinant Rabbit Monoclonal Antibody (3J2O8)
GREENER_CHOICE

Catalog No. PIMA535571
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535571 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535571 Supplier Thermo Scientific Supplier No. MA535571
Add to Cart
Add to Cart

PIAS1 Recombinant Rabbit Monoclonal Antibody (ARC1116)

PIAS1 Recombinant Monoclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen PIAS1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 3J2O8
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PIAS1
Gene Accession No. O75925, O88907
Gene Alias 2900068C24Rik; AR interacting protein; Ddxbp1; DEAD/H (Asp-Glu-Ala-Asp/His) box binding protein 1; DEAD/H box-binding protein 1; E3 SUMO-protein ligase PIAS1; E3 SUMO-protein transferase PIAS1; GBP; Gu binding protein; GU/RH-II; gu-binding protein; PIAS1; protein inhibitor of activated STAT 1; protein inhibitor of activated STAT protein 1; protein inhibitor of activated STAT, 1; protein inhibitor of activated STAT-1; RING-type E3 ubiquitin transferase PIAS1; RNA helicase II-binding protein; zinc finger, MIZ-type containing 3; ZMIZ3
Gene Symbols PIAS1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 3-101 of human PIAS1 (O75925).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56469, 8554
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.