Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIAS1/PIAS2 Recombinant Rabbit Monoclonal Antibody (8P1D2)
GREENER_CHOICE

Catalog No. PIMA538199
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538199 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538199 Supplier Invitrogen™ Supplier No. MA538199
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HepG2, Mouse brain Immunogen sequence: DNQHYNTSLL AAAAAAVSDD QDLLHSSRFF PYTSSQMFLD QLSAGGSTSL PTTNGSSSGS NSSLVSSNSL RESHSHTVTN RSSTDTASIF GIIPDIISLD.

This gene encodes a member of the protein inhibitor of activated STAT (PIAS) family. PIAS proteins function as SUMO E3 ligases and play important roles in many cellular processes by mediating the sumoylation of target proteins. This protein plays a central role as a transcriptional coregulator of numerous cellular pathways includign the STAT1 and nuclear factor kappaB pathways. Alternate splicing results in multiple transcript variants. [provided by RefSeq, Mar 2016].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIAS1/PIAS2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 8P1D2
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PIAS1
Gene Accession No. O75925, O75928, O88907, Q6AZ28, Q8C5D8
Gene Alias 2900068C24Rik; 6330408K17Rik; AI462206; androgen receptor-interacting protein 3; AR interacting protein; ARIP3; AU018068; DAB2-interacting protein; Ddxbp1; DEAD/H (Asp-Glu-Ala-Asp/His) box binding protein 1; DEAD/H box-binding protein 1; Dib; DIP; E3 SUMO-protein ligase PIAS1; E3 SUMO-protein ligase PIAS2; E3 SUMO-protein transferase PIAS1; E3 SUMO-protein transferase PIAS2; GBP; GU/RH-II; Gu-binding protein; MIZ; Miz1; msx-interacting zinc finger protein; Msx-interacting-zinc finger; msx-interacting-zinc finger protein; PIAS1; Pias2; PIAS-NY protein; PIASX; PIASxalpha; PIASX-ALPHA; PIASxalpha6; PIASxb; PIASxbeta; PIASX-BETA; protein inhibitor of activated STAT 1; protein inhibitor of activated STAT 2; protein inhibitor of activated STAT protein 1; Protein inhibitor of activated STAT x; protein inhibitor of activated STAT, 1; protein inhibitor of activated STAT, 2; protein inhibitor of activated STAT-1; Protein inhibitor of activated STAT2; RING-type E3 ubiquitin transferase PIAS1; RING-type E3 ubiquitin transferase PIAS2; RNA helicase II-binding protein; SIZ2; zinc finger, MIZ-type containing 3; zinc finger, MIZ-type containing 4; ZMIZ3; ZMIZ4
Gene Symbols PIAS1, PIAS2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 552-651 of human PIAS1+PIAS2 (O75925).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17344, 300772, 56469, 83422, 8554, 9063
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.