Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIAS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599265
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599265 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599265 Supplier Invitrogen™ Supplier No. PA599265
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PIAS1.

PIAS1 functions as an E3-type small ubiquitin-like modifier (SUMO) ligase, stabilizing the interaction between UBE2I and the substrate, and as a SUMO-tethering factor. This protein plays a crucial role in transcriptional coregulation of various cellular pathways, including the STAT pathway, the p53 pathway and the steroid hormone signaling pathway. It functions in testis as a nuclear receptor transcriptional coregulator and may have a role in androgen receptor initiation and maintenance of spermatogenesis. The effects of transcriptional coregulation, transactivation or silencing, may vary depending upon the biological context.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIAS1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PIAS1
Gene Accession No. O75925, O88907
Gene Alias 2900068C24Rik; AR interacting protein; Ddxbp1; DEAD/H (Asp-Glu-Ala-Asp/His) box binding protein 1; DEAD/H box-binding protein 1; E3 SUMO-protein ligase PIAS1; E3 SUMO-protein transferase PIAS1; GBP; Gu binding protein; GU/RH-II; gu-binding protein; PIAS1; protein inhibitor of activated STAT 1; protein inhibitor of activated STAT protein 1; protein inhibitor of activated STAT, 1; protein inhibitor of activated STAT-1; RING-type E3 ubiquitin transferase PIAS1; RNA helicase II-binding protein; zinc finger, MIZ-type containing 3; ZMIZ3
Gene Symbols PIAS1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PIAS1(Accession O75925), corresponding to amino acid residues S4-Q54.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 300772, 56469, 8554
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.