Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIAS2 Antibody (OTI2B5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246394
Description
Ensure accurate, reproducible results in Western Blot
PIAS2 Monoclonal specifically detects PIAS2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PIAS2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000 | |
Androgen receptor-interacting protein 3, ARIP3, DAB2-interacting protein, DIP, E3 SUMO-protein ligase PIAS2, MGC102682, miz, Miz1, Msx-interacting zinc finger protein, Msx-interacting-zinc finger, PIAS-NY protein, PIASX, PIASX-ALPHA, PIASX-BETA, Protein inhibitor of activated STAT x, protein inhibitor of activated STAT, 2, Protein inhibitor of activated STAT2, SIZ2, zinc finger, MIZ-type containing 4, ZMIZ4 | |
Mouse | |
68.1 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
OTI2B5 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
PIAS2 | |
Full length human recombinant protein of human PIAS2(NP_004662) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
9063 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction