Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIAS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145166
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145166 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145166 Supplier Invitrogen™ Supplier No. PA5145166
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MRPKKEAMKV SSQPCTKIES SSVLSKPCSV TVASEASKKK VDVI Antigen sequence identity: Mouse 89%; Rat 89%.

This gene encodes a protein involved in the regulation of transcription factors involved in MAP kinase signaling. The symbol MIZ1 has also been associated with ZBTB17 which is a different gene located on chromosome 1. Two alternatively spliced transcripts encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIAS2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pias2
Gene Accession No. O75928
Gene Alias 6330408K17Rik; AI462206; androgen receptor-interacting protein 3; ARIP3; AU018068; DAB2-interacting protein; Dib; DIP; E3 SUMO-protein ligase PIAS2; E3 SUMO-protein transferase PIAS2; MGC102682; MIZ; Miz1; Msx-interacting zinc finger protein; Msx-interacting-zinc finger; msx-interacting-zinc finger protein; Pias2; PIAS-NY protein; Piasx; PIASxalpha; PIASX-ALPHA; PIASxalpha6; PIASxb; PIASxbeta; PIASX-BETA; protein inhibitor of activated STAT 2; protein inhibitor of activated STAT x; protein inhibitor of activated STAT, 2; Protein inhibitor of activated STAT2; RING-type E3 ubiquitin transferase PIAS2; SIZ2; zinc finger, MIZ-type containing 4; ZMIZ4
Gene Symbols Pias2
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 427-470. Recombinant protein control fragment (Product #RP-110105).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9063
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.