Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIAS2 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519278
Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
This gene encodes a protein involved in the regulation of transcription factors involved in MAP kinase signaling. The symbol MIZ1 has also been associated with ZBTB17 which is a different gene located on chromosome 1. Two alternatively spliced transcripts encoding different isoforms have been described.
Specifications
PIAS2 | |
Polyclonal | |
Unconjugated | |
Pias2 | |
6330408K17Rik; AI462206; androgen receptor-interacting protein 3; ARIP3; AU018068; DAB2-interacting protein; Dib; DIP; E3 SUMO-protein ligase PIAS2; E3 SUMO-protein transferase PIAS2; MGC102682; MIZ; Miz1; Msx-interacting zinc finger protein; Msx-interacting-zinc finger; msx-interacting-zinc finger protein; Pias2; PIAS-NY protein; Piasx; PIASxalpha; PIASX-ALPHA; PIASxalpha6; PIASxb; PIASxbeta; PIASX-BETA; protein inhibitor of activated STAT 2; protein inhibitor of activated STAT x; protein inhibitor of activated STAT, 2; Protein inhibitor of activated STAT2; RING-type E3 ubiquitin transferase PIAS2; SIZ2; zinc finger, MIZ-type containing 4; ZMIZ4 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
9063 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O75928 | |
Pias2 | |
Synthetic peptide sequence (PQQVREICISRD) corresponding to the internal amino acids of PIAS2 (aa 185-197). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction