Learn More
PIBF Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301155AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
PIBF1 is synthesized during pregcy in response to progesterone by T lymphocytes. PIBF1 inhibits arachidonic acid release, controls NK activity, and modifies the cytokine balance exerting an anti-abortive effect. It contains a leucine zipper motif, a basic zipper sequence, a PEST sequence, a nuclear localization signal, an ER membrane retention signal and N-glycosylation and phosphorylation sites. PIBF1 is significantly higher in healthy pregt women than in women at risk for premature pregcy termination. Full-length PIBF1 is associated with the nucleus and functions as a transcription factor, whereas secretion of shorter forms which may act as cytokines is induced by activation of the cell.Specifications
| PIBF | |
| Polyclonal | |
| Unconjugated | |
| PIBF1 | |
| C13orf24, PIBF, RP11-505F3.1, progesterone-induced blocking factor 1, progesterone-induced-blocking factor 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10464 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q8WXW3 | |
| PIBF1 | |
| Between 685 and 735 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.