Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIBF Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS0420R

Description
Mediator of progesterone that by acting on the phospholipase A2 enzyme interferes with arachidonic acid metabolism, induces a Th2 biased immune response, and by controlling NK activity exerts an anti-abortive effect.Specifications
PIBF | |
Polyclonal | |
Unconjugated | |
PIBF1 | |
C13orf24; Centrosomal protein of 90 kDa; CEP90; PIBF; PIBF1; progesterone immunomodulatory binding factor 1; progesterone-induced blocking factor 1; Progesterone-induced-blocking factor 1; RGD1305077; RP11-505F3.1 | |
Rabbit | |
Protein A | |
RUO | |
10464 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q8WXW3 | |
PIBF1 | |
KLH conjugated synthetic peptide derived from human PIBF. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction