Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIBF1 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10058834DL594
Description
PIBF1 Polyclonal antibody specifically detects PIBF1 in Human samples. It is validated for Western BlotSpecifications
PIBF1 | |
Polyclonal | |
Western Blot | |
C13orf24, chromosome 13 open reading frame 24, KIAA1008, PIBF, progesterone immunomodulatory binding factor 1, progesterone-induced blocking factor 1, progesterone-induced-blocking factor 1, RP11-505F3.1 | |
The immunogen recognized by this antibody maps to a region between residue 685 and 735 of human progesterone-induced-blocking factor 1 using the numbering given in entry NP_006337.2 (GeneID 10464). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
10464 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction