Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIBF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530387
Description
Recommended positive controls: HCT116. Predicted reactivity: Human (99%), Mouse (92%), Rat (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Mediator of progesterone that by acting on the phospholipase A2 enzyme interferes with arachidonic acid metabolism, induces a Th2 biased immune response, and by controlling NK activity exerts an anti-abortive effect.
Specifications
| PIBF1 | |
| Polyclonal | |
| Unconjugated | |
| PIBF1 | |
| C13orf24; Centrosomal protein of 90 kDa; CEP90; PIBF; PIBF1; progesterone immunomodulatory binding factor 1; progesterone-induced blocking factor 1; progesterone-induced-blocking factor 1; RGD1305077; RP11-505F3.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10464 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8WXW3 | |
| PIBF1 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human PIBF1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction