Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIBF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584108
Description
Immunogen sequence: QELMKQEMET ILLRQKQLEE TNLQLREKAG DVRRNLRDFE LTEEQYIKLK AFPEDQLSIP EYVSVRFYEL VNPLRKEICE LQVKKNILAE ELSTNK.
Mediator of progesterone that by acting on the phospholipase A2 enzyme interferes with arachidonic acid metabolism, induces a Th2 biased immune response, and by controlling NK activity exerts an anti-abortive effect.
Specifications
PIBF1 | |
Polyclonal | |
Unconjugated | |
PIBF1 | |
C13orf24; Centrosomal protein of 90 kDa; CEP90; PIBF; PIBF1; progesterone immunomodulatory binding factor 1; progesterone-induced blocking factor 1; progesterone-induced-blocking factor 1; RGD1305077; RP11-505F3.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10464 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WXW3 | |
PIBF1 | |
Recombinant protein corresponding to Human PIBF1. Recombinant protein control fragment (Product #RP-104942). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction