Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIBF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA591916
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA591916 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA591916 Supplier Invitrogen™ Supplier No. PA591916
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse heart; Cellular Location: Cytoplasm, Nucleus, centriolar satellite, centrosome, cytoskeleton, microtubule organizing center Immunogen sequence: MSRKISKESK KVNISSSLES EDISLETTVP TDDISSSEER EGKVRITRQL IERKELLHNI QLLKIELSQK TMMIDNLKVD YLTKIEELEE KLNDALHQKQ.

Mediator of progesterone that by acting on the phospholipase A2 enzyme interferes with arachidonic acid metabolism, induces a Th2 biased immune response, and by controlling NK activity exerts an anti-abortive effect.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIBF1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PIBF1
Gene Accession No. Q8WXW3
Gene Alias C13orf24; Centrosomal protein of 90 kDa; CEP90; PIBF; PIBF1; progesterone immunomodulatory binding factor 1; progesterone-induced blocking factor 1; progesterone-induced-blocking factor 1; RGD1305077; RP11-505F3.1
Gene Symbols PIBF1
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10464, 306104, 52023
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.